Learn More
Eisco™ Chalcopyrite Mineral Specimen
SCI_ED
Catalog No. S27595 Shop All Eisco Products
Change view
Click to view available options
Quantity:
1/Ea.
12/Pk.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S27595 12/Pk.
S27573 1/Ea.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S27595 Supplier Eisco™ Supplier No. ESNG0066PK12
Add to Cart
Add to Cart
From the same family as fool's gold, Chalcopyrite is a yellow crystalline mineral consisting of a sulfide of copper and iron. It's the principal ore of copper formed in hydrothermal veins.
  • Correlates well to Next Generation Science Standards Evidence
  • Chalcopyrite's formation exhibits fracture and has a brassy luster which make it an excellent tool for studying economic geology
  • Great for us in comparative identification exercises
Specifications
Product Type Mineral
Grade K to 16
Quantity 12/Pk.
Specimen Size 2 to 3 cm
Weight (Metric) 0.2 kg