Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CHAMP1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA588213
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA588213 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA588213 Supplier Invitrogen™ Supplier No. PA588213
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LPKSALFSES QKAVELGDEL QIDAIDDQKC DILVQEELLA SPKKLLEDTL FPSSKKLKKD NQESSDAELS SSEYIKTDLD AMDIKGQESS SDQEQVDVES IDFSKENKMD MTSPEQSRNV LQFTEEKEAF ISEEEIAKYM KRGKGKYYCK ICCCRAMKKG AVLHHLVNKH NVHSPYKCTI CGKAFLLESL LKNHVAAHGQ SLLKCPRCNF ESNFPRGFKK HLTHCQSRHN EEANKKLMEA LEPPLEEQQI; Positive Samples: U-87MG, A-431, Jurkat ; Cellular Location: Chromosome, Cytoplasm, Nucleus, centromere, cytoskeleton, kinetochore, spindle.

Required for proper alignment of chromosomes at metaphase and their accurate segregation during mitosis. Involved in the maintenance of spindle microtubules attachment to the kinetochore during sister chromatid biorientation. May recruit CENPE and CENPF to the kinetochore.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CHAMP1
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.55 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene CHAMP1
Gene Accession No. Q96JM3
Gene Alias AA675043; AI116001; C13orf8; CAMP; CHAMP; Champ1; chromosome alignment maintaining phosphoprotein 1; chromosome alignment-maintaining phosphoprotein 1; D8Ertd457e; D8Ertd569e; Kiaa1802; mKIAA1802; MRD40; RGD1311283; Zfp828; zinc finger protein 828; ZNF828
Gene Symbols CHAMP1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 563-812 of human CHAMP1 (NP_1158121).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 283489
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.