Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Changes in Earth and Sky Curriculum Module
SCI_ED

Catalog No. S79134
Change view
Click to view available options
Mod 6 Change In Earth Sky
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
S79134 Mod 6 Change In Earth Sky
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S79134 Supplier Neo/Sci Corporation Supplier No. AZ203163

Global Changes

The surface of the Earth is effected by daily weather and rapid effects of landslides, volcanic eruptions, and earthquakes. This module will teach students why weather changes and learn how to measure these changes. Also explains the moon cycle and other objects in the sky that have patterns of movement. Comes complete with a professional development video and an assessment CD. Meets the three conceptual themes outlined in the National Science Content Standard D. Five activities. Class size: 32 students.

Earth Science Set (S79140)
Includes this curriculum module plus Properties of Earth Materials Curriculum Module and Objects in the Sky Curriculum Module.

Specifications

For Use With (Application) Can learn about weather changes, moon cycle, and other objects in the sky that have patterns of movement
Product Type Changes in Earth and Sky Curriculum Module
Sufficient For 32 Students
Includes Professional development video and assessment CD
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.