Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ Charcoal Powder
SCI_ED

Catalog No. p-4416565 Shop All Fisherbrand Products
Click to view available options
Quantity:
250 g
500 g
Packaging:
Activated Charcoal
Bone Ash
Charcoal Powder
Decolorizing Agent
This item is not returnable. View return policy
This item is not returnable. View return policy

This product is for educational use - no C of A is available.

Chemical Identifiers

CAS 7440-44-0
Molecular Formula C
Molecular Weight (g/mol) 12.011
InChI Key OKTJSMMVPCPJKN-UHFFFAOYSA-N
Synonym graphite, activated charcoal, norit, mineral, carbon-12, carbono, graphene, acticarbone, anthrasorb, carbosieve
PubChem CID 5462310
ChEBI CHEBI:27594
IUPAC Name carbon
SMILES [C]

Specifications

Color Black
Density 1.8 to 2.1g/mL
Packaging Bone Ash
Linear Formula C
Quantity 500 g
Formula Weight 12.01
Chemical Name or Material Charcoal Powder
ShelfLife 5 years
DOTInformation DOT Class 4.2, III, : Spontaneously Combustible
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.