Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Charcoal, Activated, Lab Grade, Innovating Science™

Click to view available options
Quantity:
50 g
500 g
Specifications
Specifications
Physical Form | Granular/Lump |
Quantity | 500 g |
Molecular Weight (g/mol) | 12.01 g/mol |
Percent Purity | 100% |
Grade | Laboratory |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction