Learn More
Invitrogen™ ChAT Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA577838
Description
A 1:1,000 dilution of PA5-77838 was sufficient for detection of Choline Acetyltransferase on mouse brain lysates using Goat anti-rabbit IgG:HRP as the secondary antibody. Predicted molecular weight at approximately 82.5kDa. Observed molecular weights between 68-70kDa.
ChAT catalyzes the reversible synthesis of acetylcholine (ACh) from acetyl CoA and choline at cholinergic synapses.
Specifications
ChAT | |
Polyclonal | |
Unconjugated | |
CHAT | |
acetyl CoA; acetyl CoA:choline O-acetyltransferase; B230380D24Rik; CHAT; CHOACTASE; choline acetylase; choline acetyltransferase; choline acetyltransferase (EC 2.3.1.6); choline acetyltransferase long variant; choline acetyltransferase short variant; choline O-acetyltransferase; CMS1A; CMS1A2; CMS6; LOW QUALITY PROTEIN: choline O-acetyltransferase; N-type choline acetyltransferase | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
1103, 12647 | |
-20°C | |
Liquid |
Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 50% glycerol and 0.09% sodium azide; pH 7.4 | |
P28329, Q03059 | |
CHAT | |
Synthetic peptide from the N-terminal to the mid-protein of human Choline O-Acetyltrasferase. | |
100 μg | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.