Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ChAT Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA596138
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA596138 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA596138 Supplier Invitrogen™ Supplier No. PA596138
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Samples: Raji, Mouse lung Immunogen sequence: DMYLNNRLAL PVNSSPAVIF ARQHFPGTDD QLRFAASLIS GVLSYKALLD SHSIPTDCAK GQLSGQPLCM KQYYGLFSSY RLPGHTQDTL VAQNSSIMPE.

ChAT catalyzes the reversible synthesis of acetylcholine (ACh) from acetyl CoA and choline at cholinergic synapses.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ChAT
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.39 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.3
Gene CHAT
Gene Accession No. P28329, P32738, Q03059
Gene Alias acetyl CoA; acetyl CoA:choline O-acetyltransferase; B230380D24Rik; CHAT; CHOACTASE; choline acetylase; choline acetyltransferase; choline acetyltransferase (EC 2.3.1.6); choline acetyltransferase long variant; choline acetyltransferase short variant; choline O-acetyltransferase; CMS1A; CMS1A2; CMS6; LOW QUALITY PROTEIN: choline O-acetyltransferase; N-type choline acetyltransferase
Gene Symbols CHAT
Host Species Rabbit
Immunogen Synthetic peptide.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1103, 12647, 290567
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.