Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ChAT Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA597583
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA597583 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA597583 Supplier Invitrogen™ Supplier No. PA597583
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ChAT Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), ELISA

ChAT catalyzes the reversible synthesis of acetylcholine (ACh) from acetyl CoA and choline at cholinergic synapses.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ChAT
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.8 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.5% BSA, 0.05% glycerol and 0.02% sodium azide; pH 7.4
Gene CHAT
Gene Accession No. P28329, Q03059
Gene Alias acetyl CoA; acetyl CoA:choline O-acetyltransferase; B230380D24Rik; CHAT; CHOACTASE; choline acetylase; choline acetyltransferase; choline acetyltransferase (EC 2.3.1.6); choline acetyltransferase long variant; choline acetyltransferase short variant; choline O-acetyltransferase; CMS1A; CMS1A2; CMS6; LOW QUALITY PROTEIN: choline O-acetyltransferase; N-type choline acetyltransferase
Gene Symbols CHAT
Host Species Rabbit
Immunogen Synthesized peptide derived from human Choline O-acetyltransferase protein 735-748aa.
Purification Method Antigen affinity chromatography, Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1103, 12647
Target Species Human, Mouse
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.