Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ChAT Polyclonal Antibody
GREENER_CHOICE

Catalog No. 01670566
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-670-566 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-670-566 Supplier Invitrogen™ Supplier No. PA118313
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Reconstitute in 100 μL of sterile water. Centrifuge to remove any insoluble material. After reconstitution keep aliquots at -20 °C for a higher stability, and at 4 °C with an appropriate antibacterial agent. Glycerol (1:1) may be added for an additional stability. Avoid repetitive freeze/thaw cycles.

ChAT catalyzes the reversible synthesis of acetylcholine (ACh) from acetyl CoA and choline at cholinergic synapses.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ChAT
Applications ELISA, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration Conc. Not Determined
Conjugate Unconjugated
Formulation Whole serum with no preservative
Gene CHAT
Gene Accession No. P13222, P32738, Q03059
Gene Alias acetyl CoA; acetyl CoA:choline O-acetyltransferase; B230380D24Rik; CHAT; CHOACTASE; choline acetylase; choline acetyltransferase; choline acetyltransferase (EC 2.3.1.6); choline acetyltransferase long variant; choline acetyltransferase short variant; choline O-acetyltransferase; CMS1A; CMS1A2; CMS6; LOW QUALITY PROTEIN: choline O-acetyltransferase; N-type choline acetyltransferase
Gene Symbols CHAT
Host Species Rabbit
Immunogen Synthetic peptide corresponding to residues G(167) L F S S Y R L P G H T Q D T L V A Q K S S(188) of porcine ChAT conjugated to KLH.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100342226, 12647, 290567, 396896
Target Species Mouse, Rat, Rabbit, Pig
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.