Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CHCHD1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA544059
Description
Peptide sequence: MATPSLRGRL ARFGNPRKPV LKPNKPLILA NRVGERRREK GEATCITEMS Sequence homology: Cow: 86%; Dog: 86%; Goat: 86%; Guinea Pig: 79%; Horse: 83%; Human: 100%; Mouse: 79%; Rat: 79%.
CHCHD1 (Coiled-Coil-Helix-Coiled-Coil-Helix Domain Containing 1) is a Protein Coding gene. Among its related pathways are Mitochondrial translation and Organelle biogenesis and maintenance.
Specifications
CHCHD1 | |
Polyclonal | |
Unconjugated | |
Chchd1 | |
1110001O19Rik; 2400010G13Rik; 28S ribosomal protein S37, mitochondrial; C10orf34; C2360; CHCHD1; coiled-coil-helix-coiled-coil-helix domain containing 1; coiled-coil-helix-coiled-coil-helix domain-containing protein 1; Mitochondrial small ribosomal subunit protein mS37; MRPS37; MRP-S37; Nuclear protein C2360; similar to coiled-coil-helix-coiled-coil-helix domain containing 1 | |
Rabbit | |
Affinity chromatography | |
RUO | |
118487 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q96BP2 | |
Chchd1 | |
Synthetic peptide directed towards the N-terminal of human CHCHD1 (aa 1-50). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction