Learn More
Invitrogen™ CHCHD1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5114012
Description
Immunogen sequence: KEIQGFLDCA ARAQEARKMR SIQETLGESG SLLPNKLNKL LQRFPNKPYL For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles. Predicted homology: Cow: 93%; Dog: 90%; Goat: 86%; Guinea Pig: 100%; Horse: 86%; Human: 100%; Mouse: 83%; Rat: 93%.
CHCHD1 (Coiled-Coil-Helix-Coiled-Coil-Helix Domain Containing 1) is a Protein Coding gene. Among its related pathways are Mitochondrial translation and Organelle biogenesis and maintenance.
Specifications
| CHCHD1 | |
| Polyclonal | |
| Unconjugated | |
| Chchd1 | |
| 1110001O19Rik; 2400010G13Rik; 28S ribosomal protein S37, mitochondrial; C10orf34; C2360; CHCHD1; coiled-coil-helix-coiled-coil-helix domain containing 1; coiled-coil-helix-coiled-coil-helix domain-containing protein 1; Mitochondrial small ribosomal subunit protein mS37; MRPS37; MRP-S37; Nuclear protein C2360; similar to coiled-coil-helix-coiled-coil-helix domain containing 1 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 118487 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q96BP2 | |
| Chchd1 | |
| Synthetic peptide directed towards the middle region of human CHCHD1. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.