Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CHCHD10 Monoclonal Antibody (OTI3D7), TrueMAB™, OriGene

Mouse Monoclonal Antibody
Supplier: OriGene TA811808

Description
CHCHD10 encodes a mitochondrial protein that is enriched at cristae junctions in the intermembrane space. It may play a role in cristae morphology maintece or oxidative phosphorylation. Mutations in this gene cause frontotemporal dementia and/or amyotrophic lateral sclerosis-2. Alternative splicing of this gene results in multiple transcript variants. Related pseudogenes have been identified on chromosomes 7 and 19.Specifications
CHCHD10 | |
Monoclonal | |
1 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
Q8WYQ3 | |
CHCHD10 | |
Human recombit protein fragment corresponding to amino acids 17-142 of human CHCHD10 produced in E.coli. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
Western Blot | |
OTI3D7 | |
Unconjugated | |
CHCHD10 | |
C22orf16, FTDALS2, IMMD, N27C7-4, SMAJ | |
Mouse | |
Affinity Chromatography | |
RUO | |
400916 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction