Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CHCHD10 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5100435
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5100435 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5100435 Supplier Invitrogen™ Supplier No. PA5100435
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total CHCHD10.

CHCHD10 encodes a mitochondrial protein that is enriched at cristae junctions in the intermembrane space. It may play a role in cristae morphology maintenance or oxidative phosphorylation. Mutations in this gene cause frontotemporal dementia and/or amyotrophic lateral sclerosis-2. Alternative splicing of this gene results in multiple transcript variants. Related pseudogenes have been identified on chromosomes 7 and 19.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CHCHD10
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene CHCHD10
Gene Accession No. Q8WYQ3
Gene Alias 1620401E04Rik; AI267078; C22orf16; CHCHD10; coiled-coil-helix-coiled-coil-helix domain containing 10; coiled-coil-helix-coiled-coil-helix domain-containing protein 10, mitochondrial; FTDALS2; IMMD; N27C7-4; Ndg2; Nur77 downstream gene 2; Nur77 downstream protein 2; Protein N27C7-4; SMAJ
Gene Symbols CHCHD10
Host Species Rabbit
Immunogen A synthesized peptide derived from human CHCHD10(Accession Q8WYQ3), corresponding to amino acid residues P94-P142.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 103172, 400916
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.