Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CHCHD10 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA551935
Description
Immunogen sequence: GLMAQMATTA AGVAVGSAVG HVMGSALTGA FSGGSSEPSQ PAVQQAPTPA APQPLQMGPC AYEIRQFLDC STTQSDLSLC EGFSEALKQC KYYHGLSSLP Highest antigen sequence identity to the following orthologs: Mouse - 87%, Rat - 89%.
CHCHD10 encodes a mitochondrial protein that is enriched at cristae junctions in the intermembrane space. It may play a role in cristae morphology maintenance or oxidative phosphorylation. Mutations in this gene cause frontotemporal dementia and/or amyotrophic lateral sclerosis-2. Alternative splicing of this gene results in multiple transcript variants. Related pseudogenes have been identified on chromosomes 7 and 19.
Specifications
| CHCHD10 | |
| Polyclonal | |
| Unconjugated | |
| CHCHD10 | |
| 1620401E04Rik; AI267078; C22orf16; CHCHD10; coiled-coil-helix-coiled-coil-helix domain containing 10; coiled-coil-helix-coiled-coil-helix domain-containing protein 10, mitochondrial; FTDALS2; IMMD; N27C7-4; Ndg2; Nur77 downstream gene 2; Nur77 downstream protein 2; Protein N27C7-4; SMAJ | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 400916 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q8WYQ3 | |
| CHCHD10 | |
| Recombinant protein corresponding to Human CHCHD10. Recombinant protein control fragment (Product #RP-91517). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction