Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CHCHD2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA565727
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA565727 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA565727 Supplier Invitrogen™ Supplier No. PA565727
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PRGSRSRTSRM APPASRAPQM RAAPRPA Highest antigen sequence identity to the following orthologs - mouse 89%, rat 86%.

Transcription factor. Binds to the oxygen responsive element of COX4I2 and activates its transcription under hypoxia conditions (4% oxygen), as well as normoxia conditions (20% oxygen).
TRUSTED_SUSTAINABILITY

Specifications

Antigen CHCHD2
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.10 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CHCHD2
Gene Accession No. Q9Y6H1
Gene Alias 16.7kD protein; AAG10; Aging-associated gene 10 protein; AL033347; C7orf17; CHCHD2; coiled-coil-helix-coiled-coil-helix domain containing 2; coiled-coil-helix-coiled-coil-helix domain-containing protein 2; coiled-coil-helix-coiled-coil-helix domain-containing protein 2, mitochondrial; ethanol induced 6; Etohi6; HCV NS2 trans-regulated protein; mitochondria nuclear retrograde regulator 1; MNRR1; NS2TP; PARK22
Gene Symbols CHCHD2
Host Species Rabbit
Immunogen Recombinant Human CHCHD2. Recombinant protein control fragment (Product #RP-104947).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51142
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.