Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CHCHD2 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501987246
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-7246 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-7246 Supplier Bioss Supplier No. BS6407R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Transcription factor. Binds to the oxygen responsive element of COX4I2 and activates its transcription under hypoxia conditions (4% oxygen), as well as normoxia conditions (20% oxygen).

Specifications

Antigen CHCHD2
Applications Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene CHCHD2
Gene Accession No. Q9Y6H1
Gene Alias 16.7kD protein; AAG10; Aging-associated gene 10 protein; AL033347; C7orf17; CHCHD2; coiled-coil-helix-coiled-coil-helix domain containing 2; Coiled-coil-helix-coiled-coil-helix domain-containing protein 2; coiled-coil-helix-coiled-coil-helix domain-containing protein 2, mitochondrial; ethanol induced 6; Etohi6; HCV NS2 trans-regulated protein; mitochondria nuclear retrograde regulator 1; MNRR1; NS2TP; PARK22
Gene Symbols CHCHD2
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human CHCHD2 (96-130aa).
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51142
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.