Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CHCHD3 Antibody (OTI7G4), Alexa Fluor™ 405, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP271975AF405
Description
CHCHD3 Monoclonal specifically detects CHCHD3 in Human, Mouse, Rat samples. It is validated for Western Blot.Specifications
CHCHD3 | |
Monoclonal | |
Alexa Fluor 405 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
CHCHD3 | |
Recombinant protein fragment corresponding to amino acids 2-227 of human CHCHD3 (NP_060282) produced in E.coli. | |
0.1 mL | |
Primary | |
Human, Mouse, Rat | |
IgG1 |
Western Blot | |
OTI7G4 | |
Western Blot | |
coiled-coil-helix-coiled-coil-helix domain containing 3, FLJ20420, mitochondrial | |
Mouse | |
Affinity Purified | |
metabolism | |
54927.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction