Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CHCHD3 Antibody (OTI7G4) - Azide and BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP271975
Description
CHCHD3 Monoclonal specifically detects CHCHD3 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.Specifications
CHCHD3 | |
Monoclonal | |
LYOPH | |
Western Blot 1:2000 | |
coiled-coil-helix-coiled-coil-helix domain containing 3, FLJ20420, mitochondrial | |
Mouse | |
Affinity Purified | |
metabolism | |
54927 | |
Human, Mouse, Rat | |
Lyophilized |
Western Blot | |
OTI7G4 | |
Unconjugated | |
Lyophilized from PBS (pH 7.3) with 8% Trehalose with No Preservative | |
CHCHD3 | |
Human recombinant protein fragment corresponding to amino acids 2-227 of human CHCHD3 (NP_060282) produced in E.coli. | |
100 μg | |
Primary | |
Reconstitute with PBS (pH 7.3). To use this carrier-free antibody for conjugation experiments, another round of desalting is highly recommended. | |
Store at -20°C. Avoid freeze-thaw cycles. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction