Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CHCHD3 Monoclonal Antibody (OTI3E10), TrueMAB™, OriGene
SDP

Catalog No. 501679814
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-167-9814 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-167-9814 Supplier OriGene Supplier No. TA803543
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Component of the MICOS complex, a large protein complex of the mitochondrial inner membrane that plays crucial roles in the maintece of crista junctions, inner membrane architecture, and formation of contact sites to the outer membrane. Has also been shown to function as a transcription factor which binds to the BAG1 promoter and represses BAG1 transcription. Plays an important role in the maintece of the MICOS complex stability and the mitochondrial cristae morphology.

Specifications

Antigen CHCHD3
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI3E10
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene CHCHD3
Gene Accession No. Q9NX63
Gene Alias MINOS3, Mic19, PPP1R22
Gene Symbols CHCHD3
Host Species Mouse
Immunogen Human recombit protein fragment corresponding to amino acids 2-227 of human CHCHD3 produced in E.coli.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54927
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.