Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CHCHD3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA597124
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA597124 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA597124 Supplier Invitrogen™ Supplier No. PA597124
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MGGTTSTRRV TFEADENENI TVVKGIRLSE NVIDRMKESS PSGSKSQRYS GAYGASVSDE ELKRRVAEEL ALEQAKKESE DQKRLKQAKE LDRERAAANE QLTRAILRER ICSEEERAKA KHLARQLEEK DRVLKKQDAF YKEQLARLEE RSSEFYRVTT EQYQKAAEEV EAKFKRYESH PVCADLQAKI LQCYRENTHQ TLKCSALATQ YMHCVNHAKQ SMLEKGG; Positive Samples: A-549, HeLa, A-431, Jurkat, HepG2, mouse kidney, mouse liver, rat heart, rat kidney, Mouse kidney, Mouse liver, Mouse heart; Cellular Location: Cytoplasm, Intermembrane side, Lipid-anchor, Mitochondrion, Mitochondrion inner membrane, Nucleus.

Component of the MICOS complex, a large protein complex of the mitochondrial inner membrane that plays crucial roles in the maintenance of crista junctions, inner membrane architecture, and formation of contact sites to the outer membrane. Has also been shown to function as a transcription factor which binds to the BAG1 promoter and represses BAG1 transcription. Plays an important role in the maintenance of the MICOS complex stability and the mitochondrial cristae morphology.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CHCHD3
Applications ELISA, Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.61 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene CHCHD3
Gene Accession No. Q9CRB9, Q9NX63
Gene Alias 0610041L09Rik; 1700039J09Rik; AW558177; CHCHD3; coiled-coil-helix-coiled-coil-helix domain containing 3; Coiled-coil-helix-coiled-coil-helix domain-containing protein 3; coiled-coil-helix-coiled-coil-helix domain-containing protein 3, mitochondrial; FLJ20420; Mic19; MICOS complex subunit MIC19; MINOS3; mitochondrial inner membrane organizing system 3; PPP1R22; protein phosphatase 1, regulatory subunit 22
Gene Symbols CHCHD3
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-227 of human CHCHD3 (NP_0602821).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 296966, 54927, 66075
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.