Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CHCHD4 Monoclonal Antibody (6C9)
GREENER_CHOICE

Catalog No. PIMA524519
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA524519 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA524519 Supplier Invitrogen™ Supplier No. MA524519
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Peptide Sequence: MSYCRQEGKD RIIFVTKEDH ETPSSAELVA DDPNDPYEEH GLILPNGNIN WNCPCLGGMA SGPCGEQFKS AFSCFHYSTE EIKGSDCVDQ FRAMQECMQK YPDLYPQEDE DEEEEREKKP AEQAEETAPI EATATKEEEG SS.

CHCHD4, a component of human mitochondria, belongs to a protein family whose members share 6 highly conserved cysteine residues constituting a -CXC-CX(9)C-CX(9)C- motif in the C terminus (Hofmann et al., 2005).
TRUSTED_SUSTAINABILITY

Specifications

Antigen CHCHD4
Applications ELISA, Western Blot
Classification Monoclonal
Clone 6C9
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Gene CHCHD4
Gene Accession No. Q8N4Q1
Gene Alias 2410012P20Rik; 2810014D17Rik; AI838740; CHCHD4; coiled-coil-helix-coiled-coil-helix domain containing 4; coiled-coil-helix-coiled-coil-helix domain-containing protein 4; MIA40; mitochondrial intermembrane space import and assembly 40 homolog; mitochondrial intermembrane space import and assembly protein 40; TIMM40; translocase of inner mitochondrial membrane 40 homolog
Gene Symbols CHCHD4
Host Species Mouse
Immunogen CHCHD4 (AAH33775.1, 1 a.a. ~ 142 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 131474
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.