Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CHCHD4 Polyclonal Antibody
CHCHD4 Polyclonal Antibody
Supplier: Thermo Scientific PA589016
Description
CHCHD4 Polyclonal Antibody for Western Blot
Specifications
| CHCHD4 | |
| Polyclonal | |
| Unconjugated | |
| CHCHD4 | |
| 2410012P20Rik; 2810014D17Rik; AI838740; CHCHD4; coiled-coil-helix-coiled-coil-helix domain containing 4; coiled-coil-helix-coiled-coil-helix domain-containing protein 4; MIA40; mitochondrial intermembrane space import and assembly 40 homolog; mitochondrial intermembrane space import and assembly protein 40; TIMM40; translocase of inner mitochondrial membrane 40 homolog | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 131474, 312559, 72170 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| 1.71 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| Q8N4Q1, Q8VEA4 | |
| CHCHD4 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-142 of human CHCHD4 (NP_0010919721). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction