Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CHCHD5 Monoclonal Antibody (OTI3H3), TrueMAB™, OriGene

Mouse Monoclonal Antibody
Supplier: OriGene TA502399S


Description
CHCHD5 Monoclonal antibody specifically detects CHCHD5 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), Western BlotSpecifications
CHCHD5 | |
Monoclonal | |
0.35 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
Q8WV99 | |
Mouse | |
Affinity chromatography | |
RUO | |
130617 | |
-20° C, Avoid Freeze/Thaw Cycles | |
IgG1 |
Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
OTI3H3 | |
Unconjugated | |
ZFAND2B | |
1110027L01Rik; AW045710; C2orf9; Chchd5; coiled-coil-helix-coiled-coil-helix domain containing 5; coiled-coil-helix-coiled-coil-helix domain-containing protein 5; MIC14; mitochondrial intermembrane space cysteine motif protein of 14 kDa homolog | |
Full length human recombinant protein of human ZFAND2B produced in HEK293T cell. | |
30 μL | |
Primary | |
Human | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction