Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CHCHD5 Monoclonal Antibody (OTI6G5), TrueMAB™, OriGene
SDP

Catalog No. 502541592
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
30 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-1592 30 μL
50-167-3435 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-254-1592 Supplier OriGene Supplier No. TA502374S
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CHCHD5 Monoclonal antibody specifically detects CHCHD5 in Human samples. It is validated for Flow Cytometry, Western Blot

Specifications

Antigen CHCHD5
Applications Flow Cytometry, Western Blot
Classification Monoclonal
Clone OTI6G5
Concentration 0.41 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene CHCHD5
Gene Accession No. Q9BSY4
Gene Alias 1110027L01Rik; AW045710; C2orf9; Chchd5; coiled-coil-helix-coiled-coil-helix domain containing 5; coiled-coil-helix-coiled-coil-helix domain-containing protein 5; MIC14; mitochondrial intermembrane space cysteine motif protein of 14 kDa homolog
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 1-110 of human CHCHD5 produced in E.coli.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84269
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.