Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CHCHD6 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA534486
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA534486 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA534486 Supplier Invitrogen™ Supplier No. PA534486
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

A suggested positive control is SK-N-SH cell lysate. PA5-34486 can be used with blocking peptide PEP-1528.

CHCHD6 is a member of a family of proteins containing a conserved (coiled coil 1)-(helix 1)-(coiled coil 2)-(helix 2) domain and has been observed in a complex with the mitochondrial proteins mitofilin, SAM50, metaxins 1 and 2, and CHCHD3. CHCHD6 knockdown causes severe defects in mitochondrial cristae morphology as well as reductions in cell growth, ATP production and oxygen consumption. This decrease of CHCHD6 also led to a reduction in mitofilin protein levels, while a knockdown of mitofilin resulted in a reduced level of CHCHD6, suggesting coordinate regulation between these two proteins. CHCHD6 knockdown in human cancer cells enhances their sensitivity to genotoxic anticancer drugs, suggesting that CHCHD6 may be a potential therapeutic anti-tumor target.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CHCHD6
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene CHCHD6
Gene Accession No. Q9BRQ6
Gene Alias 0710001P09Rik; 1700021B03Rik; CHCHD6; CHCM1; coiled coil helix cristae morphology 1; coiled coil helix cristae morphology protein 1; coiled-coil-helix cristae morphology 1; coiled-coil-helix cristae morphology protein 1; coiled-coil-helix-coiled-coil-helix domain containing 6; Coiled-coil-helix-coiled-coil-helix domain-containing protein 6; coiled-coil-helix-coiled-coil-helix domain-containing protein 6, mitochondrial; Mic25; MICOS complex subunit MIC25; PPP1R23; protein phosphatase 1, regulatory subunit 23
Gene Symbols CHCHD6
Host Species Rabbit
Immunogen A 17 amino acid peptide near the amino terminus of human CHCHD6.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84303
Target Species Human
Content And Storage Maintain refrigerated at 2°C to 8°C for up to 3 months. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.