Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CHCHD7 Antibody, Biotin, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP281709B
This item is not returnable.
View return policy
Description
CHCHD7 Polyclonal antibody specifically detects CHCHD7 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISASpecifications
CHCHD7 | |
Polyclonal | |
PBS | |
Rabbit | |
Peptide affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Western Blot, ELISA | |
Biotin | |
coiled-coil-helix-coiled-coil-helix domain containing 7, coiled-coil-helix-coiled-coil-helix domain-containing protein 7, FLJ40966, MGC2217 | |
Rabbit polyclonal CHCHD7 antibody was raised against a 15 amino acid peptide near the carboxy terminus of human CHCHD7. The immunogen is located within the last 50 amino acids of CHCHD7. Amino Acid Squence: NSIVMQRRKNGVKPF | |
0.1 mL | |
Cancer, Cell Cycle and Replication | |
79145 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction