Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CHCHD7 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA562241
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA562241 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA562241 Supplier Invitrogen™ Supplier No. PA562241
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: EETHAPNSNW VYVMLPSKKT VRMPSVTQRL RDPDINPCLS ESDASTRCLD ENNYDRERCS TYFLRYKNCR RFWNSIVMQR RKNGVKPFMP Highest antigen sequence identity to the following orthologs: Mouse - 63%, Rat - 63%.

CHCHD7 is a newly identified member of a multifamily of proteins containing a conserved (coiled coil 1)-(helix 1)-(coiled coil 2)-(helix 2) domain. Northern blot analysis revealed that the gene is ubiquitously expressed. Its biological function is unknown and the gene has hitherto not been associated with neoplasia. CHCHD7 and PLAG1 are located head-to-head about 500 bp apart in 8q12. A chromosomal aberration involving CHCHD7 is found in salivary gland pleiomorphic adenomas, the most common benign epithelial tumors of the salivary gland.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CHCHD7
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CHCHD7
Gene Accession No. Q9BUK0
Gene Alias 1810049H20Rik; CHCHD7; coiled-coil-helix-coiled-coil-helix domain containing 7; coiled-coil-helix-coiled-coil-helix domain-containing protein 7; COX23; COX23 cytochrome c oxidase assembly homolog
Gene Symbols CHCHD7
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human CHCHD7 (23-85). Recombinant protein control fragment (Product #RP-93898).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79145
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.