Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CHCHD7 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5116845
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5116845 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5116845 Supplier Invitrogen™ Supplier No. PA5116845
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Samples: Mouse kidney Immunogen sequence: MHQTRTGKKT VRMPSVTQRL RDPDINPCLS ESDASTRCLD ENNYDRERCS TYFLRYKNCR RFWNSIVMQR RKNGVKPFMP TAAERDEILR AVGNMPY.

CHCHD7 is a newly identified member of a multifamily of proteins containing a conserved (coiled coil 1)-(helix 1)-(coiled coil 2)-(helix 2) domain. Northern blot analysis revealed that the gene is ubiquitously expressed. Its biological function is unknown and the gene has hitherto not been associated with neoplasia. CHCHD7 and PLAG1 are located head-to-head about 500 bp apart in 8q12. A chromosomal aberration involving CHCHD7 is found in salivary gland pleiomorphic adenomas, the most common benign epithelial tumors of the salivary gland.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CHCHD7
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 4.27 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene CHCHD7
Gene Accession No. Q8K2Q5, Q9BUK0
Gene Alias 1810049H20Rik; CHCHD7; coiled-coil-helix-coiled-coil-helix domain containing 7; coiled-coil-helix-coiled-coil-helix domain-containing protein 7; COX23; COX23 cytochrome c oxidase assembly homolog
Gene Symbols CHCHD7
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-97 of human CHCHD7 (NP_0772762).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 66433, 684258, 79145
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.