Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CHD-8 Mouse anti-Human, Clone: 7F4.1, MilliporeSigma™

Catalog No. MABN2265 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABN2265 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABN2265 Supplier MilliporeSigma Supplier No. MABN2265
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-CHD-8, clone 7F4.1 is a highly specific mouse monoclonal antibody that targets Chromodomain-helicase-DNA-binding protein 8 and has been tested for use in Immunohistochemistry (Paraffin) and Western Blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CHD-8
Applications Western Blot, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 7F4.1
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Gene Symbols CHD8,HELSNF1,KIAA1564
Host Species Mouse
Immunogen GST/His-tagged recombinant fragment corresponding to 112 amino acids rom the C-terminal half of human Chromodomain-helicase-DNA-binding protein 8. The immunogen sequence is conserved in both isoforms.
Purification Method Protein G Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) NP_001164100
Test Specificity Clone 7F4.1 detects human Chromodomain-helicase-DNA-binding protein 8 (CHD8). It targets an epitope within 112 amino acids from the C-terminal half.
Target Species Human
Content And Storage Stable for 1 year at 2-8°C from date of receipt.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.