Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CHD1 Antibody, Allophycocyanin, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP298902APC
Description
CHD1 Polyclonal antibody specifically detects CHD1 in Human, Mouse samples. It is validated for Immunohistochemistry (Paraffin)Specifications
CHD1 | |
Polyclonal | |
Immunohistochemistry-Paraffin | |
ATP-dependent helicase CHD1, CHD-1, chromodomain helicase DNA binding protein 1, chromodomain-helicase-DNA-binding protein 1, DKFZp686E2337, EC 3.6.1, EC 3.6.4.12 | |
Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of the Human CHD1. | |
0.1 mL | |
Chromatin Modifiers | |
1105 | |
Store at 4C in the dark. | |
IgG |
Immunohistochemistry (Paraffin) | |
APC | |
PBS | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Human, Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction