Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CHD1 Monoclonal Antibody (2F11), Active Motif™
SDP

Catalog No. 501993019
Encompass
Change view
Click to view available options
Quantity:
10 μg
100 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-199-3019 100 μg
50-199-3020 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-199-3019 Supplier Active Motif Supplier No. 39989
Add to Cart
Add to Cart

Rat Monoclonal Antibody

CHD1 is an ATP-dependent chromatin remodeling factor, containing two tandem chromodomains. In human both chromodomains are essential for specific binding to histone H3, methylated at lysine 4. CHD1 belongs to the SNF2/RAD54 helicase family and contains also one helicase ATP-binding domain and one helicase C-terminal domain. It may play an important role in gene regulation.

Specifications

Antigen CHD1
Applications Immunocytochemistry, Immunofluorescence, Western Blot
Classification Monoclonal
Clone 2F11
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 70mM tris with 0.07mM EDTA, 105mM NaCl, 30% glycerol, 31mM glycine and 0.035% sodium azide; pH 8
Gene CHD1
Gene Accession No. O14646, P40201
Gene Alias 4930525N21Rik; AI851787; ATP-dependent helicase CHD1; AW555109; Chd1; Chd-1; chromodomain helicase DNA binding protein 1; Chromodomain-helicase-DNA-binding protein 1; DKFZp686E2337; EC 3.6.1; EC 3.6.4.12; LOW QUALITY PROTEIN: chromodomain-helicase-DNA-binding protein 1
Gene Symbols CHD1
Host Species Rat
Immunogen This CHD1 antibody was raised against a peptide corresponding to the C-terminus of mouse CHD1.
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1105, 12648
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.