Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CHD1 Mouse anti-Human, Clone: 43133, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP266776
Description
CHD1 Monoclonal specifically detects CHD1 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
CHD1 | |
Monoclonal | |
2 mg/mL | |
Western Blot 1:100-1:2000, Immunohistochemistry 1:10 - 1:500, Immunohistochemistry-Paraffin | |
ATP-dependent helicase CHD1, CHD-1, chromodomain helicase DNA binding protein 1, chromodomain-helicase-DNA-binding protein 1, DKFZp686E2337, EC 3.6.1, EC 3.6.4.12 | |
Mouse | |
Protein A purified | |
Primary | |
Human, Mouse | |
Purified |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
43133 | |
Unconjugated | |
PBS (pH7.4), 0.2% BSA, 40% Glycerol with 0.05% Sodium Azide | |
CHD1 | |
Recombinant protein within Human CHD1 aa 1,479-1,710/ 1,710. (SwissProt: O14646 Human) | |
100 μL | |
1105 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction