Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CHD1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA582795
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA582795 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA582795 Supplier Invitrogen™ Supplier No. PA582795
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: QESQQNSDQN SNLNPHVIRN PDVERLKENT NHDDSSRDSY SSDRHLTQYH DHHKDRHQGD SYKKSDSRKR PYSSFSNGKD HRDWDHYKQD SRYYSDREKH RKLDDHRSRD HRSNLEGSL Highest antigen sequence indentity to the following orthologs: Mouse - 90%, Rat - 88%.

CHD1 is an ATP-dependent chromatin remodeling factor, containing two tandem chromodomains. In human both chromodomains are essential for specific binding to histone H3, methylated at lysine 4. CHD1 belongs to the SNF2/RAD54 helicase family and contains also one helicase ATP-binding domain and one helicase C-terminal domain. It may play an important role in gene regulation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CHD1
Applications ChIP Assay, Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CHD1
Gene Accession No. O14646
Gene Alias 4930525N21Rik; AI851787; ATP-dependent helicase CHD1; AW555109; Chd1; Chd-1; chromodomain helicase DNA binding protein 1; chromodomain-helicase-DNA-binding protein 1; DKFZp686E2337; EC 3.6.1; EC 3.6.4.12; LOW QUALITY PROTEIN: chromodomain-helicase-DNA-binding protein 1
Gene Symbols CHD1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human CHD1. Recombinant protein control fragment (Product #RP-91650).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1105
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.