Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CHD1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5121150
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5121150 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5121150 Supplier Invitrogen™ Supplier No. PA5121150
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Positive test controls include: Jurkat. The target is usually found in the following locations: Cytoplasm, Nucleus. Immunogen sequence: DREKHRKLDD HRSRDHRSNL EGSLKDRSHS DHRSHSDHRL HSDHRSSSEY THHKSSRDYR YHSDWQMDHR ASSSGPRSPL DQRSPYGSRS PFEHSVEHKS TPEHTWSSRK T.

CHD1 is an ATP-dependent chromatin remodeling factor, containing two tandem chromodomains. In human both chromodomains are essential for specific binding to histone H3, methylated at lysine 4. CHD1 belongs to the SNF2/RAD54 helicase family and contains also one helicase ATP-binding domain and one helicase C-terminal domain. It may play an important role in gene regulation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CHD1
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.39 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene CHD1
Gene Accession No. O14646, P40201
Gene Alias 4930525N21Rik; AI851787; ATP-dependent helicase CHD1; AW555109; Chd1; Chd-1; chromodomain helicase DNA binding protein 1; chromodomain-helicase-DNA-binding protein 1; DKFZp686E2337; EC 3.6.1; EC 3.6.4.12; LOW QUALITY PROTEIN: chromodomain-helicase-DNA-binding protein 1
Gene Symbols CHD1
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 1600 to the C-terminus of human CHD1 (NP_001261.2).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1105, 12648, 308215
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.