Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CHD1 Polyclonal Antibody, Active Motif™
SDP

Catalog No. 501994106
Encompass
Change view
Click to view available options
Quantity:
10 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-199-4106 10 μL
50-199-4105 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-199-4106 Supplier Active Motif Supplier No. 39730
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

CHD1 is an ATP-dependent chromatin remodeling factor, containing two tandem chromodomains. In human both chromodomains are essential for specific binding to histone H3, methylated at lysine 4. CHD1 belongs to the SNF2/RAD54 helicase family and contains also one helicase ATP-binding domain and one helicase C-terminal domain. It may play an important role in gene regulation.

Specifications

Antigen CHD1
Applications Western Blot
Classification Polyclonal
Concentration Conc. Not Determined
Conjugate Unconjugated
Formulation whole serum with 30% glycerol and 0.035% sodium azide; pH
Gene CHD1
Gene Accession No. O14646, P40201
Gene Alias 4930525N21Rik; AI851787; ATP-dependent helicase CHD1; AW555109; Chd1; Chd-1; chromodomain helicase DNA binding protein 1; Chromodomain-helicase-DNA-binding protein 1; DKFZp686E2337; EC 3.6.1; EC 3.6.4.12; LOW QUALITY PROTEIN: chromodomain-helicase-DNA-binding protein 1
Gene Symbols CHD1
Host Species Rabbit
Immunogen This CHD1 antibody was raised against a recombinant protein corresponding to amino acids 251-467 of human CHD1.
Quantity 10 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1105, 12648
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.