Learn More
CHD1 Rabbit anti-Human, Polyclonal, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A301217A

Description
The recommended shelf life for this product is 1 year from date of receipt.
The CHD family of proteins is characterized by the presence of chromo (chromatin organization modifier) domains and SNF2-related helicase/ATPase domains. CHD genes alter gene expression possibly by modification of chromatin structure.Specifications
CHD1 | |
Polyclonal | |
Unconjugated | |
CHD1 | |
CHD-1, ATPase DNA-binding protein 1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
1105 | |
4° C | |
Liquid |
Immunoprecipitation | |
1 mg/ml | |
phosphate, tris citrate with 0.09% sodium azide; pH 7-8 | |
O14646 | |
CHD1 | |
Between 1500 and 1550 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.