Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CHD1, Rabbit anti-Human, Polyclonal Antibody, Abnova™

Catalog No. 89998414 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-998-414 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-998-414 Supplier Abnova Corporation Supplier No. PAB31299
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Specifications

Antigen CHD1
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot (1:500) The optimal working dilution should be determined by the end user.
Formulation In PBS (0.05% sodium azide, 0.05% proclin 300).
Gene chromodomain helicase DNA binding protein 1
Gene Alias DKFZp686E2337
Gene Symbols CHD1
Host Species Rabbit
Immunogen Recombinant GST fusion protein corresponding to human CHD1.
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1105
Target Species Human
Content And Storage Store at -20°C. For long term storage store at -80°C. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.