Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CHD1L Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA598591
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA598591 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA598591 Supplier Invitrogen™ Supplier No. PA598591
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

CHD1L Polyclonal Antibody for Western Blot, ICC/IF, ELISA

In response to DNA strand breaks, chromatin adopts a relaxed structure due to the addition of poly(ADP-ribose) (PAR) to chromatin proteins by PARP enzymes (see PARP1; MIM 173870), and this relaxation facilitates the repair of DNA damage. CHD1L interacts with PAR and has a role in chromatin relaxation following DNA damage (Ahel et al., 2009 [PubMed 19661379]).
TRUSTED_SUSTAINABILITY

Specifications

Antigen CHD1L
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 3.86 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene CHD1L
Gene Accession No. Q86WJ1
Gene Alias 4432404A22Rik; ALC1; amplified in liver cancer 1; amplified in liver cancer protein 1; CHD1L; CHDL; chromodomain helicase DNA binding protein 1 like; chromodomain helicase DNA binding protein 1-like; chromodomain-helicase-DNA-binding protein 1-like; LOW QUALITY PROTEIN: chromodomain-helicase-DNA-binding protein 1-like; Snf2p; Unknown (protein for MGC:128792)
Gene Symbols CHD1L
Host Species Rabbit
Immunogen Recombinant Human Chromodomain-helicase-DNA-binding protein 1-like protein (704-897AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9557
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.