Learn More
Invitrogen™ CHD2 Polyclonal Antibody

Description
Immunogen sequence: QHWYKDHHYG DRRHMDAHRS GSYRPNNMSR KRPYDQYSSD RDHRGHRDYY DRHHHDSKRR RSDEFRPQNY HQQDFRRMS Highest antigen sequence indentity to the following orthologs: Rat - 96%, Mouse - 96%.
Specifications
Specifications
| Antigen | CHD2 |
| Applications | Immunohistochemistry (Paraffin), Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | CHD2 |
| Gene Accession No. | O14647 |
| Gene Alias | 2810013C04Rik; 2810040A01Rik; 5630401D06Rik; AI851092; ATP-dependent helicase CHD2; BC029703; chd2; CHD-2; chromodomain helicase DNA binding protein 2; chromodomain-helicase-DNA-binding protein 2; DKFZp547I1315; DKFZp686E01200; DKFZp781D1727; Down syndrome cell adhesion molecule; EEOC; FLJ38614; LOW QUALITY PROTEIN: chromodomain-helicase-DNA-binding protein 2; si:ch211-196l7.5 |
| Gene Symbols | CHD2 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.