Learn More
Invitrogen™ CHD2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA579039
Description
Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: monkey COS-7 whole cell, human Siha whole cell, rat brain tissue, rat C6 whole cell, mouse brain tissue, mouse NIH/3T3 whole cell. IHC: rat brain tissue, mouse brain tissue.
The CHD family of proteins is characterized by the presence of chromo domains and SNF2-related helicase/ATPase domains. CHD genes alter gene expression possibly by modification of chromatin structure thus altering access of the transcriptional apparatus to its chromosomal DNA template. Alternatively spliced transcript variants encoding distinct isoforms have been found for this gene.
Specifications
CHD2 | |
Polyclonal | |
Unconjugated | |
CHD2 | |
2810013C04Rik; 2810040A01Rik; 5630401D06Rik; AI851092; ATP-dependent helicase CHD2; BC029703; chd2; CHD-2; chromodomain helicase DNA binding protein 2; chromodomain-helicase-DNA-binding protein 2; DKFZp547I1315; DKFZp686E01200; DKFZp781D1727; Down syndrome cell adhesion molecule; EEOC; FLJ38614; LOW QUALITY PROTEIN: chromodomain-helicase-DNA-binding protein 2; si:ch211-196l7.5 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
1106, 244059, 308738 | |
-20°C | |
Lyophilized |
Immunohistochemistry (Paraffin), Western Blot | |
500 μg/mL | |
PBS with 5 mg BSA and 0.05 mg sodium azide | |
E9PZM4, O14647 | |
CHD2 | |
E. coli-derived human CHD2 recombinant protein (Position: R1124-K1351). | |
100 μg | |
Primary | |
Human, Mouse, Rat, Monkey | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.