Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CHD3 Antibody, Janelia Fluor™ 646, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB10060412JF646
Description
CHD3 Polyclonal antibody specifically detects CHD3 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
CHD3 | |
Polyclonal | |
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin | |
ATP-dependent helicase CHD3, CHD-3, chromodomain helicase DNA binding protein 3, chromodomain-helicase-DNA-binding protein 3, EC 3.6.1, EC 3.6.4.12, hZFH, Mi-2 autoantigen 240 kDa protein, Mi-2a, Mi2-ALPHA, ZFH, Zinc finger helicase, zinc-finger helicase (Snf2-like) | |
The immunogen recognized by this antibody maps to a region between residue 1575 and 1625 of human chromodomain helicase DNA binding protein 3 using the numbering given in entry NP_001005273.1 (GeneID 1107). | |
0.1 mL | |
Immune Dysfunction, Immune System Diseases, Immunology, modENCODE Antibodies | |
1107 | |
Store at 4C in the dark. | |
IgG |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Janelia Fluor 646 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction