Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CHD3 Recombinant Mouse Monoclonal Antibody, AbFlex™, Active Motif™
SDP

Catalog No. 501993794
Encompass
Change view
Click to view available options
Quantity:
10 μg
100 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-199-3794 10 μg
50-199-3793 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-199-3794 Supplier Active Motif Supplier No. 91274
Add to Cart
Add to Cart

Mouse Recombinant Monoclonal Antibody

This gene encodes a member of the CHD family of proteins which are characterized by the presence of chromo domains and SNF2-related helicase/ATPase domains. This protein is one of the components of a histone deacetylase complex referred to as the Mi-2/NuRD complex which participates in the remodeling of chromatin by deacetylating histones. Chromatin remodeling is essential for many processes including transcription. Autoantibodies against this protein are found in a subset of patients with dermatomyositis. Three alternatively spliced transcripts encoding different isoforms have been described.

Specifications

Antigen CHD3
Applications Immunoprecipitation, Western Blot
Classification Monoclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 140mM HEPES, 32mM sodium acetate with 30% glycerol, 70mM NaCl and 0.035% sodium azide; pH 7.5
Gene CHD3
Gene Accession No. O16102
Gene Alias 2600010P09Rik; AF020312; ATP-dependent helicase CHD3; CHD3; CHD-3; Chd7; chromodomain helicase DNA binding protein 3; Chromodomain-helicase-DNA-binding protein 3; hZFH; MGC40857; Mi 2a; Mi2 ALPHA; Mi-2 alpha; Mi-2 autoantigen 240 kDa protein; Mi-2a; Mi2-alpha; Prp7; Prp9 1; Prp9-1; ZFH; Zinc finger helicase; zinc-finger helicase (Snf2-like)
Gene Symbols CHD3
Host Species Mouse
Immunogen This antibody was raised against a synthetic peptide corresponding to amino acid residues 865-885 of Drosophila.
Purification Method Protein A
Quantity 10 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 40111
Target Species Drosophila
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.