Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CHD3 Recombinant Rabbit Monoclonal Antibody (JE57-05)
Rabbit Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA544909
This item is not returnable.
View return policy
Description
CHD3 Recombinant Monoclonal Antibody for Western Blot, IHC (P), Flow
This gene encodes a member of the CHD family of proteins which are characterized by the presence of chromo domains and SNF2-related helicase/ATPase domains. This protein is one of the components of a histone deacetylase complex referred to as the Mi-2/NuRD complex which participates in the remodeling of chromatin by deacetylating histones. Chromatin remodeling is essential for many processes including transcription. Autoantibodies against this protein are found in a subset of patients with dermatomyositis. Three alternatively spliced transcripts encoding different isoforms have been described.
Specifications
CHD3 | |
Recombinant Monoclonal | |
1 mg/mL | |
TBS with 0.05% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4 | |
Q12873 | |
CHD3 | |
Recombinant protein within human CHD3 aa 1,850-2,000. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
JE57-05 | |
Unconjugated | |
CHD3 | |
2600010P09Rik; AF020312; ATP-dependent helicase CHD3; CG9594; CG9594-PA; CHD3; CHD-3; Chd3-PA; Chd7; chromodomain helicase DNA binding protein 3; chromodomain-helicase-DNA-binding protein 3; dCHD3; DmCHD3; Dmel\CG9594; Dmel_CG9594; hZFH; MGC40857; Mi 2a; Mi2 ALPHA; Mi-2 alpha; mi-2 autoantigen 240 kDa protein; Mi-2a; Mi2-alpha; Prp7; Prp9 1; Prp9-1; ZFH; Zinc finger helicase; zinc-finger helicase (Snf2-like) | |
Rabbit | |
Protein A | |
RUO | |
1107 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction