Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CHD4 Antibody, Allophycocyanin, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB10057519APC
Description
CHD4 Polyclonal antibody specifically detects CHD4 in Human, Mouse samples. It is validated for Western Blot, Immunoprecipitation, ChIP assay SequencingSpecifications
CHD4 | |
Polyclonal | |
Western Blot, Immunoprecipitation, Chromatin Immunoprecipitation Sequencing | |
ATP-dependent helicase CHD4, CHD-4, chromodomain helicase DNA binding protein 4, chromodomain-helicase-DNA-binding protein 4, DKFZp686E06161, EC 3.6.1, EC 3.6.4.12, Mi-2 autoantigen 218 kDa protein, Mi-2b, Mi2-BETA | |
Maps to a region between residue 25 and 75 of human chromodomain helicase DNA binding protein 4 (Mi2 autoantigen 218 kDa protein) using the numbering given in entry NP_001264.2 (GeneID 1108). | |
0.1 mL | |
Immune System Diseases, Immunology | |
1108 | |
Store at 4C in the dark. | |
IgG |
Western Blot, Immunoprecipitation, ChIP sequencing (ChIP-seq) | |
APC | |
PBS | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human, Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction