Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CHD4 Antibody, Biotin, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP299098B
Description
CHD4 Polyclonal specifically detects CHD4 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
CHD4 | |
Polyclonal | |
Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin | |
ATP-dependent helicase CHD4, CHD-4, chromodomain helicase DNA binding protein 4, chromodomain-helicase-DNA-binding protein 4, DKFZp686E06161, EC 3.6.1, EC 3.6.4.12, Mi-2 autoantigen 218 kDa protein, Mi-2b, Mi2-BETA | |
Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of human CHD4. | |
0.1 mL | |
Immune System Diseases, Immunology | |
1108 | |
Store at 4°C in the dark. | |
IgG |
Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Biotin | |
PBS | |
Rabbit | |
Antigen Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction