Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CHD4 Antibody (CHD4 3F2/4), mFluor Violet 500 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP250163MFV500
Description
CHD4 Monoclonal antibody specifically detects CHD4 in Human, Mouse samples. It is validated for Flow CytometrySpecifications
CHD4 | |
Monoclonal | |
mFluor Violet 500 SE | |
50mM Sodium Borate | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Human, Mouse | |
Purified |
Flow Cytometry | |
CHD4 3F2/4 | |
Flow Cytometry | |
ATP-dependent helicase CHD4, CHD-4, chromodomain helicase DNA binding protein 4, chromodomain-helicase-DNA-binding protein 4, DKFZp686E06161, EC 3.6.1, EC 3.6.4.12, Mi-2 autoantigen 218 kDa protein, Mi-2b, Mi2-BETA | |
Synthetic peptide (N- and C- terminal peptides were used for immunisation. 1. ASGLGSPSPCSAGSEEEDM & 2. CSRLANRAPEPPPQQVAQQQ. Which peptide the antibody recognises has not been tested) | |
0.1 mL | |
Immune System Diseases, Immunology | |
1108 | |
Store at 4C in the dark. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction