Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CHD4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA532181
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA532181 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA532181 Supplier Invitrogen™ Supplier No. PA532181
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: 293T, A431, HeLa, HepG2. Predicted reactivity: Mouse (100%), Xenopus laevis (82%), Bovine (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

The product of this gene belongs to the SNF2/RAD54 helicase family. It represents the main component of the nucleosome remodeling and deacetylase complex and plays an important role in epigenetic transcriptional repression. Patients with dermatomyositis develop antibodies against this protein.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CHD4
Applications Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7
Gene CHD4
Gene Accession No. Q14839, Q6PDQ2
Gene Alias 9530019N15Rik; AA617397; ATP-dependent helicase CHD4; BC005710; Chd4; CHD-4; chromodomain helicase DNA binding protein 4; chromodomain-helicase-DNA-binding protein 4; D6Ertd380e; Mi-2; Mi-2 autoantigen; Mi-2 autoantigen 218 kDa protein; Mi-2 beta; MI2B; Mi-2b; Mi-2beta; Mi2-BETA; mKIAA4075
Gene Symbols CHD4
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human CHD4. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 107932, 1108
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.