Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CHD4 Recombinant Rabbit Monoclonal Antibody (BLR066G)
Rabbit Recombinant Monoclonal Antibody
$173.10 - $436.00
Specifications
Antigen | CHD4 |
---|---|
Clone | BLR066G |
Concentration | 40 μg/mL |
Content And Storage | 4°C |
Applications | Flow Cytometry, Immunoprecipitation, Western Blot, Western Blot |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
PIMA544323
|
Invitrogen™
MA544323 |
100 μL |
Each for $436.00
|
|
|||||
PIMA544324
|
Invitrogen™
MA544324 |
20 μL |
Each for $173.10
|
|
|||||
Description
CHD4 Recombinant Monoclonal Antibody for Western Blot, Flow, IP
The product of this gene belongs to the SNF2/RAD54 helicase family. It represents the main component of the nucleosome remodeling and deacetylase complex and plays an important role in epigenetic transcriptional repression. Patients with dermatomyositis develop antibodies against this protein.Specifications
CHD4 | |
40 μg/mL | |
Flow Cytometry, Immunoprecipitation, Western Blot, Western Blot | |
Unconjugated | |
Rabbit | |
RUO | |
BBS with 0.1% BSA and 0.09% sodium azide; pH 7.8-8.6 | |
Q14839 | |
1108 | |
Peptide representing a region between residue 25 and 75 of human Chromodomain-helicase-DNA-binding protein 4. | |
Antibody |
BLR066G | |
4°C | |
Recombinant Monoclonal | |
Liquid | |
IgG | |
Human | |
CHD4 | |
9530019N15Rik; AA617397; ATP-dependent helicase CHD4; BC005710; Chd4; CHD-4; chromodomain helicase DNA binding protein 4; chromodomain-helicase-DNA-binding protein 4; D6Ertd380e; Mi-2; Mi-2 autoantigen; Mi-2 autoantigen 218 kDa protein; Mi-2 beta; MI2B; Mi-2b; Mi-2beta; Mi2-BETA; mKIAA4075 | |
CHD4 | |
Primary | |
Purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title