Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CHD6 Antibody, DyLight 350, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB10060414UV
Description
CHD6 Polyclonal antibody specifically detects CHD6 in Human samples. It is validated for Western BlotSpecifications
CHD6 | |
Polyclonal | |
Western Blot | |
ATP-dependent helicase CHD6, CHD5dJ620E11.1, CHD-6, chromodomain helicase DNA binding protein 6, EC 3.6.1, EC 3.6.4.12, FLJ22369, KIAA1335chromodomain-helicase-DNA-binding protein 6, Radiation-induced gene B protein, RIGBhelicase C-terminal domain- and SNF2 N-terminal domain-containing protein | |
The immunogen recognized by this antibody maps to a region between residue 2665 and 2715 of human chromodomain helicase DNA binding protein 6 using the numbering given in entry NP_115597.3 (GeneID 84181). | |
0.1 mL | |
Primary | |
Human | |
Purified |
Western Blot | |
DyLight 350 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
84181 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction