Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CHD6 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5103740
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5103740 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5103740 Supplier Invitrogen™ Supplier No. PA5103740
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total CHD6.

This gene encodes a member of the SNF2/RAD54 helicase protein family. The encoded protein contains two chromodomains, a helicase domain, and an ATPase domain. Several multi-subunit protein complexes remodel chromatin to allow patterns of cell type-specific gene expression, and the encoded protein is thought to be a core member of one or more of these chromatin remodeling complexes. The encoded protein may function as a transcriptional repressor and is involved in the cellular repression of influenza virus replication.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CHD6
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene CHD6
Gene Accession No. A3KFM7, Q8TD26
Gene Alias 5430439G14Rik; 6330406J24Rik; ATP-dependent helicase CHD6; CHD5; Chd6; CHD-6; chromodomain helicase DNA binding protein 6; chromodomain-helicase-DNA-binding protein 6; helicase C-terminal domain- and SNF2 N-terminal domain-containing protein; Kiaa1335; Radiation-induced gene B protein; RIGB
Gene Symbols CHD6
Host Species Rabbit
Immunogen A synthesized peptide derived from human CHD6(Accession Q8TD26), corresponding to amino acid residues S2645-E2695.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 71389, 84181
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.